Assay Details
| Gene Symbol: INSM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101230.1 | NP_001094700.1 | 1 | 2148 | 137 | |
| GenBank mRNA | BC146266.1 | - | 1 | 2148 | 137 | |
More Information
Set Membership: |
|

INSM1
Genomic Map