Assay Details
| Gene Symbol: C23H6orf47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101267.1 | NP_001094737.1 | 1 | 1349 | 93 | |
| GenBank mRNA | BC150103.1 | - | 1 | 1349 | 93 | |
More Information
Set Membership: |
|

C23H6orf47
Genomic Map