Assay Details
| Gene Symbol: RAB12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102292.1 | NP_001095762.1 | 6 | 1749 | 141 | |
| GenBank mRNA | BC149542.1 | - | 6 | 1749 | 141 | |
More Information
Set Membership: |
|

RAB12
Genomic Map