Assay Details
| Gene Symbol: PLAGL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103289.1 | NP_001096759.1 | 6 | 1849 | 151 | |
| GenBank mRNA | BC149832.1 | - | 6 | 1849 | 151 | |
More Information
Set Membership: |
|

PLAGL1
Genomic Map