Assay Details
| Gene Symbol: PLCXD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103304.2 | NP_001096774.1 | 3 | 1650 | 134 | |
| GenBank mRNA | BC149012.1 | - | 3 | 1651 | 134 | |
More Information
Set Membership: |
|

PLCXD3
Genomic Map