Assay Details
| Gene Symbol: CENPW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111261.1 | NP_001104731.1 | 3 | 441 | 147 | |
| GenBank mRNA | BC148115.1 | - | 3 | 441 | 147 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CENPW| Gene Symbol: CENPW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111261.1 | NP_001104731.1 | 3 | 441 | 147 | |
| GenBank mRNA | BC148115.1 | - | 3 | 441 | 147 | |
Set Membership: |
|