Assay Details
| Gene Symbol: MRPL33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113308.1 | NP_001106779.1 | 1 | 349 | 95 | |
| GenBank mRNA | BC103150.1 | - | 1 | 349 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MRPL33| Gene Symbol: MRPL33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113308.1 | NP_001106779.1 | 1 | 349 | 95 | |
| GenBank mRNA | BC103150.1 | - | 1 | 349 | 95 | |
Set Membership: |
|