Assay Details
| Gene Symbol: PRM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174157.4 | NP_776582.2 | 1 | 255 | 164 | |
| GenBank mRNA | BC109783.1 | - | 1 | 258 | 164 | |
| X16559.1 | - | 255 | 164 | |||
More Information
Set Membership: |
|

PRM2
Genomic Map