Assay Details
| Gene Symbol: YWHAH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174492.2 | NP_776917.2 | 2 | 749 | 127 | |
| GenBank mRNA | BC102982.1 | - | 2 | 749 | 127 | |
| J03868.1 | - | 2 | 748 | 127 | ||
More Information
Set Membership: |
|

YWHAH
Genomic Map