Assay Details
Gene Symbol: ADIPOQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174742.2 | NP_777167.1 | 3 | 755 | 121 | |
GenBank mRNA | AF269230.1 | - | 3 | 755 | 121 | |
BC140488.1 | - | 2 | 708 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
