Assay Details
| Gene Symbol: BASP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174780.3 | NP_777205.1 | 2 | 1448 | 88 | |
| GenBank mRNA | BC123462.1 | - | 2 | 1448 | 88 | |
| U92535.1 | - | 2 | 1310 | 88 | ||
More Information
Set Membership: |
|

BASP1
Genomic Map