Assay Details
| Gene Symbol: YWHAG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174793.2 | NP_777218.2 | 2 | 1352 | 128 | |
| GenBank mRNA | BC153255.1 | - | 2 | 1352 | 128 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

YWHAG| Gene Symbol: YWHAG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174793.2 | NP_777218.2 | 2 | 1352 | 128 | |
| GenBank mRNA | BC153255.1 | - | 2 | 1352 | 128 | |
Set Membership: |
|