Assay Details
| Gene Symbol: PROK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199445.1 | NP_955516.1 | 2 | 153 | 67 | |
| GenBank mRNA | AY192557.1 | - | 2 | 153 | 67 | |
| AY192558.1 | - | 2 | 153 | 67 | ||
More Information
Set Membership: |
|

PROK2
Genomic Map