Assay Details
| Gene Symbol: LYZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080339.1 | NP_001073808.1 | 4 | 554 | 160 | |
| GenBank mRNA | M26245.1 | - | 4 | 554 | 160 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LYZ1| Gene Symbol: LYZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080339.1 | NP_001073808.1 | 4 | 554 | 160 | |
| GenBank mRNA | M26245.1 | - | 4 | 554 | 160 | |
Set Membership: |
|