Assay Details
| Gene Symbol: PNN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242585.1 | NP_001229514.1 | 9 | 1483 | 175 | |
| GenBank mRNA | BC103403.1 | - | 9 | 1467 | 175 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: PNN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242585.1 | NP_001229514.1 | 9 | 1483 | 175 | |
| GenBank mRNA | BC103403.1 | - | 9 | 1467 | 175 | |
Set Membership: |
|