Assay Details
| Gene Symbol: LYZ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180999.1 | NP_851342.1 | 1 | 55 | 89 | |
| GenBank mRNA | L23758.1 | - | 1 | 141 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_180999.1 transcripts.
Set Membership: |
|

LYZ2
Genomic Map