Assay Details
| Gene Symbol: RAB3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046606.2 | NP_001040071.1 | 5 | 1953 | 144 | |
| GenBank mRNA | BC112795.1 | - | 5 | 1954 | 144 | |
More Information
Set Membership: |
|

RAB3C
Genomic Map