Assay Details
| Gene Symbol: IFITM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078141.2 | NP_001071609.1 | 1 | 48 | 75 | |
| GenBank mRNA | BC103086.1 | - | 1 | 49 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC103086.1, NM_001078141.2 transcripts.
Set Membership: |
|

IFITM3
Genomic Map