Assay Details
| Gene Symbol: RNASE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014386.4 | NP_001014408.2 | 2 | 749 | 145 | |
| GenBank mRNA | BC149636.1 | - | 2 | 750 | 145 | |
| S80747.1 | - | 1 | 622 | 145 | ||
More Information
Set Membership: |
|

RNASE1
Genomic Map