Assay Details
| Gene Symbol: PCDHGC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103334.1 | NP_001096804.1 | 1 | 55 | 112 | |
| NM_001114080.1 | NP_001107552.1 | 1 | 71 | 112 | ||
| GenBank mRNA | BC140535.1 | - | 1 | 71 | 112 | |
| BC149092.1 | - | 1 | 55 | 112 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC140535.1, BC149092.1, NM_001103334.1, NM_001114080.1 transcripts.
Set Membership: |
|

PCDHGC3
Genomic Map