Assay Details
| Gene Symbol: SPINK2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114858.2 | NP_001108330.1 | 3 - 4 | 239 | 91 | |
| GenBank mRNA | BC109719.1 | - | 3 - 4 | 240 | 91 | |
More Information
Set Membership: |
|

SPINK2B
Genomic Map