Assay Details
| Gene Symbol: SRP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115083.2 | NP_001108555.1 | 1 - 2 | 184 | 118 | |
| GenBank mRNA | BC111640.1 | - | 1 - 2 | 185 | 118 | |
More Information
Set Membership: |
|

SRP9
Genomic Map