Assay Details
| Gene Symbol: CDX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206299.1 | NP_001193228.1 | 1 - 2 | 690 | 79 | |
| GenBank mRNA | AM293662.1 | - | 1 - 2 | 52 | 79 | |
More Information
Set Membership: |
|

CDX2
Genomic Map