Assay Details
| Gene Symbol: H2AC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205596.1 | NP_001192525.1 | 1 | 113 | 59 | |
| NM_001355022.1 | NP_001341951.1 | 1 | 113 | 59 | ||
| GenBank mRNA | DQ066891.1 | - | 1 | 55 | 59 | |
More Information
Set Membership: |
|

H2AC6
Genomic Map