Assay Details
| Gene Symbol: HACD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130757.2 | NP_001124229.1 | 1 - 2 | 234 | 65 | |
| GenBank mRNA | BC112557.1 | - | 1 - 2 | 236 | 65 | |
More Information
Set Membership: |
|

HACD2
Genomic Map