Assay Details
| Gene Symbol: ATOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130758.1 | NP_001124230.1 | 3 - 4 | 300 | 90 | |
| GenBank mRNA | BC102437.1 | - | 3 - 4 | 300 | 90 | |
More Information
Set Membership: |
|

ATOX1
Genomic Map