Assay Details
| Gene Symbol: CHCHD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130759.2 | NP_001124231.1 | 2 - 3 | 120 | 66 | |
| GenBank mRNA | BC118483.1 | - | 2 - 3 | 121 | 66 | |
More Information
Set Membership: |
|

CHCHD7
Genomic Map