Assay Details
| Gene Symbol: CALCB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143737.2 | NP_001137209.1 | 3 - 4 | 299 | 67 | |
| GenBank mRNA | AB462435.1 | - | 3 - 4 | 300 | 67 | |
More Information
Set Membership: |
|

CALCB
Genomic Map