Assay Details
| Gene Symbol: PGD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143738.1 | NP_001137210.1 | 8 - 9 | 934 | 82 | |
| GenBank mRNA | BC102178.1 | - | 8 - 9 | 934 | 82 | |
More Information
Set Membership: |
|

PGD
Genomic Map