Assay Details
| Gene Symbol: TOMM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143740.1 | NP_001137212.1 | 1 - 2 | 174 | 90 | |
| GenBank mRNA | BC108221.1 | - | 1 - 2 | 174 | 90 | |
More Information
Set Membership: |
|

TOMM5
Genomic Map