Assay Details
| Gene Symbol: ZNF438 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143772.3 | NP_001137244.1 | 7 - 8 | 2183 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF438| Gene Symbol: ZNF438 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143772.3 | NP_001137244.1 | 7 - 8 | 2183 | 83 | |
Set Membership: |
|