Assay Details
| Gene Symbol: ABL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143855.2 | NP_001137327.1 | 10 - 11 | 1850 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ABL2| Gene Symbol: ABL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143855.2 | NP_001137327.1 | 10 - 11 | 1850 | 63 | |
Set Membership: |
|