Assay Details
| Gene Symbol: SCN2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144109.3 | NP_001137581.1 | 11 - 12 | 1955 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SCN2A| Gene Symbol: SCN2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144109.3 | NP_001137581.1 | 11 - 12 | 1955 | 66 | |
Set Membership: |
|