Assay Details
| Gene Symbol: CFAP73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144873.1 | NP_001138345.1 | 1 - 2 | 171 | 86 | |
| GenBank mRNA | EE906766.1 | - | 1 - 2 | 136 | 86 | |
| EV638271.1 | - | 1 - 2 | 174 | 86 | ||
More Information
Set Membership: |
|

CFAP73
Genomic Map