Assay Details
| Gene Symbol: CFAP73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144873.1 | NP_001138345.1 | 6 - 7 | 979 | 77 | |
| GenBank mRNA | CB166585.1 | - | 2 - 3 | 160 | 77 | |
More Information
Set Membership: |
|

CFAP73
Genomic Map