Assay Details
| Gene Symbol: TBL1X | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145232.1 | NP_001138704.1 | 11 - 12 | 1291 | 54 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TBL1X| Gene Symbol: TBL1X | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145232.1 | NP_001138704.1 | 11 - 12 | 1291 | 54 | |
Set Membership: |
|