Assay Details
| Gene Symbol: HCAR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145234.1 | NP_001138706.1 | 1 | 850 | 96 | |
| GenBank mRNA | EU809465.1 | - | 1 | 850 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HCAR1| Gene Symbol: HCAR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145234.1 | NP_001138706.1 | 1 | 850 | 96 | |
| GenBank mRNA | EU809465.1 | - | 1 | 850 | 96 | |
Set Membership: |
|