Assay Details
Gene Symbol: C23H6orf141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145653.1 | NP_001139125.1 | 1 - 2 | 901 | 102 | |
GenBank mRNA | DT825475.1 | - | 1 - 2 | 314 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
