Assay Details
| Gene Symbol: CRH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013400.2 | NP_001013418.2 | 2 | 619 | 88 | |
| GenBank mRNA | DV822182.1 | - | 3 | 473 | 88 | |
| EE338630.1 | - | 3 | 473 | 88 | ||
More Information
Set Membership: |
|

CRH
Genomic Map