Assay Details
| Gene Symbol: C3H1orf226 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083790.2 | NP_001077259.1 | 1 - 2 | 799 | 98 | |
| GenBank mRNA | BC134601.1 | - | 799 | 98 | ||
More Information
Set Membership: |
|

C3H1orf226
Genomic Map