Assay Details
| Gene Symbol: CKAP2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098901.2 | NP_001092371.1 | 1 - 2 | 73 | 64 | |
| GenBank mRNA | BC142323.1 | - | 1 - 2 | 74 | 64 | |
More Information
Set Membership: |
|

CKAP2L
Genomic Map