Assay Details
| Gene Symbol: MARS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098971.1 | NP_001092441.1 | 1 | 2154 | 97 | |
| GenBank mRNA | BC146211.1 | - | 1 | 2154 | 97 | |
| BT026161.1 | - | 1 | 2137 | 97 | ||
More Information
Set Membership: |
|

MARS2
Genomic Map