Assay Details
| Gene Symbol: COX8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114517.2 | NP_001107989.1 | 1 - 2 | 121 | 63 | |
| GenBank mRNA | J05202.1 | - | 1 - 2 | 121 | 63 | |
More Information
Set Membership: |
|

COX8B
Genomic Map