Assay Details
| Gene Symbol: MIC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127317.1 | NP_001120789.1 | 5 - 6 | 842 | 115 | |
| GenBank mRNA | EU645577.1 | - | 5 - 6 | 842 | 115 | |
| Gene Symbol: MIC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281907.1 | NP_001268836.1 | 5 - 6 | 115 | ||
More Information
Set Membership: |
|

MIC1
Genomic Map