Assay Details
| Gene Symbol: PFN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128197.1 | NP_001121669.1 | 1 - 2 | 152 | 88 | |
| GenBank mRNA | BC122616.1 | - | 1 - 2 | 152 | 88 | |
More Information
Set Membership: |
|

PFN2
Genomic Map