Assay Details
| Gene Symbol: CHMP4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128504.2 | NP_001121976.1 | 1 - 2 | 114 | 65 | |
| GenBank mRNA | BC133471.1 | - | 1 - 2 | 115 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC133471.1, NM_001128504.2 transcripts.
Set Membership: |
|

CHMP4A
Genomic Map