Assay Details
| Gene Symbol: CHMP4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128506.1 | NP_001121978.1 | 2 - 3 | 499 | 79 | |
| GenBank mRNA | BC123447.1 | - | 2 - 3 | 499 | 79 | |
More Information
Set Membership: |
|

CHMP4B
Genomic Map