Assay Details
| Gene Symbol: KRT15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130747.1 | NP_001124219.1 | 7 - 8 | 1333 | 88 | |
| GenBank mRNA | BC118284.1 | - | 7 - 8 | 1333 | 88 | |
More Information
Set Membership: |
|

KRT15
Genomic Map