Assay Details
| Gene Symbol: GALNTL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076441.1 | NP_001069909.1 | 9 - 10 | 1475 | 72 | |
| GenBank mRNA | BC111260.1 | - | 9 - 10 | 1475 | 72 | |
| KT763324.1 | - | 8 - 9 | 1354 | 72 | ||
More Information
Set Membership: |
|

GALNTL5
Genomic Map