Assay Details
| Gene Symbol: MAGEA11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080732.2 | NP_001074201.1 | 1 - 2 | 117 | 80 | |
| GenBank mRNA | BC122817.1 | - | 1 - 2 | 119 | 80 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC122817.1, NM_001080732.2 transcripts.
Set Membership: |
|

MAGEA11
Genomic Map